Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABARAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007278.1 | NP_009209.1 | 4 | 440 | ||
GenBank mRNA | AB030711.1 | - | 4 | 439 | ||
AF044671.1 | - | 4 | 414 | |||
AF067171.1 | - | 4 | 440 | |||
AF161586.1 | - | 4 | 404 | |||
AF183425.1 | - | 4 | 439 | |||
AK098634.1 | - | 4 | 444 | |||
AK312205.1 | - | 4 | 435 | |||
AL050182.1 | - | 346 | ||||
BC106748.1 | - | 4 | 382 | 40022383 | ||
BC106749.1 | - | 4 | 382 | 40022384 | ||
CR457043.1 | - | 4 | 336 | |||
CR542235.1 | - | 4 | 336 | |||
U65413.1 | - | 28 |