Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: U2AF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012478.1 | NP_001012496.1 | 2, 3 | 1250 | ||
NM_007279.2 | NP_009210.1 | 2, 3 | 1250 | |||
XM_006722994.2 | XP_006723057.1 | 3, 4 | 1288 | |||
XM_011526410.1 | XP_011524712.1 | 3, 4 | 1288 | |||
GenBank mRNA | AB451261.1 | - | 2, 3 | 195 | ||
AB451387.1 | - | 2, 3 | 195 | |||
BC008740.2 | - | 2, 3 | 254 | 3346087 | ||
BC030574.2 | - | 2, 3 | 1154 | 3687471 | ||
X64044.1 | - | 2, 3 | 235 |