Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLCA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012128.3 | NP_036260.2 | 8 | 1387 | ||
NR_024602.1 | 7 | 1278 | ||||
XM_011541015.2 | XP_011539317.1 | 8 | 1206 | |||
GenBank mRNA | AF127035.1 | - | 8 | 1371 | ||
AK000072.1 | - | 8 | 1385 | |||
AK000138.1 | - | 1 | 64 | |||
AK293766.1 | - | 7 | 1042 | |||
AK307785.1 | - | 7 | 1223 | |||
AK307898.1 | - | 6 | 992 | |||
AK314430.1 | - | 8 | 1383 | |||
AY358470.1 | - | 8 | 1367 | |||
BC113687.1 | - | 8 | 1382 | 8322739 | ||
BC113689.1 | - | 8 | 1381 | 8322741 |
Gene Symbol: LOC105378828 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135837.1 | 5 | 1460 | |||
GenBank mRNA | AI346357.1 | - | 1 | 57 |