Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBOX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267584.1 | NP_001254513.1 | 3 | 1413 | ||
NM_014948.3 | NP_055763.1 | 3 | 1413 | |||
NM_199415.2 | NP_955447.1 | 3 | 1413 | |||
GenBank mRNA | AB020667.1 | - | 3 | 1381 | ||
AK022444.1 | - | 3 | 1377 | |||
AK222570.1 | - | 3 | 1385 | |||
AK222876.1 | - | 3 | 1338 | |||
BC000515.2 | - | 3 | 1333 | 2822816 | ||
BC046122.1 | - | 3 | 1373 | 5554044 | ||
CR457089.1 | - | 2 | 1227 |