Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301132.1 | NP_001288061.1 | 2 | 521 | ||
NM_014952.4 | NP_055767.3 | 2 | 394 | |||
XM_011521366.2 | XP_011519668.2 | 2 | 518 | |||
XM_011521367.2 | XP_011519669.2 | 2 | 523 | |||
XM_011521368.2 | XP_011519670.2 | 2 | 517 | |||
XM_011521369.2 | XP_011519671.1 | 2 | 372 | |||
GenBank mRNA | AB023162.1 | - | 2 | 521 | ||
BC022782.2 | - | 2 | 333 | 3903581 |