Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAXIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007349.3 | NP_031375.3 | 12 | 2365 | ||
XM_005249539.1 | XP_005249596.1 | 13 | 2504 | |||
XM_011515982.2 | XP_011514284.1 | 12 | 2479 | |||
XM_011515983.1 | XP_011514285.1 | 12 | 2365 | |||
XM_017011881.1 | XP_016867370.1 | 13 | 3677 | |||
GenBank mRNA | AK093584.1 | - | 6 | 1058 | ||
AK123600.1 | - | 13 | 2504 | |||
AK126674.1 | - | 4 | 2373 | |||
AK293744.1 | - | 10 | 2197 | |||
BC008328.1 | - | 3 | 289 | |||
BC033781.1 | - | 7 | 1606 | 4509264 | ||
BX538201.1 | - | 6 | 1141 | |||
BX640616.1 | - | 12 | 4263 | |||
U80735.1 | - | 8 | 1348 |