Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SORCS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014978.2 | NP_055793.1 | 3 | 1002 | ||
GenBank mRNA | AB028982.1 | - | 3 | 1002 | ||
BC117417.1 | - | 3 | 818 | 40125968 |