Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012308.2 | NP_036440.1 | 2 | 878 | ||
NR_027473.1 | 2 | 878 | ||||
XM_011544860.2 | XP_011543162.1 | 2 | 937 | |||
XM_017017391.1 | XP_016872880.1 | 1 | 62 | |||
GenBank mRNA | AB023221.2 | - | 2 | 195 | ||
AK024505.1 | - | 2 | 120 | |||
BC047486.1 | - | 2 | 880 | 5296445 | ||
BC064360.1 | - | 2 | 293 | 5534384 | ||
JQ710741.1 | - | 1 | 62 | |||
JQ710742.1 | - | 1 | 62 | |||
JQ710743.1 | - | 1 | 62 | |||
JQ710744.1 | - | 1 | 62 |