Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MON2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278469.1 | NP_001265398.1 | 30 | 4792 | ||
NM_001278470.1 | NP_001265399.1 | 29 | 4774 | |||
NM_001278471.1 | NP_001265400.1 | 29 | 4705 | |||
NM_001278472.1 | NP_001265401.1 | 31 | 4870 | |||
NM_015026.2 | NP_055841.2 | 30 | 4792 | |||
XM_017019041.1 | XP_016874530.1 | 31 | 4795 | |||
XM_017019042.1 | XP_016874531.1 | 30 | 4777 | |||
XM_017019043.1 | XP_016874532.1 | 32 | 4869 | |||
XM_017019044.1 | XP_016874533.1 | 32 | 4871 | |||
XM_017019045.1 | XP_016874534.1 | 33 | 4539 | |||
XM_017019046.1 | XP_016874535.1 | 27 | 3964 | |||
GenBank mRNA | AB017814.1 | - | 30 | 4517 | ||
AB028963.3 | - | 31 | 4763 | |||
AK022671.1 | - | 4 | 969 | |||
AK092646.1 | - | 4 | 1556 | |||
AL833066.2 | - | 30 | 4787 | |||
AL834320.1 | - | 24 | 3750 | |||
BC136621.1 | - | 29 | 4743 | 9020611 | ||
BC144309.1 | - | 29 | 4674 | 9052829 | ||
BC151241.1 | - | 31 | 4786 | 8860514 | ||
BX537415.1 | - | 30 | 4790 |