Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZHX3 SIRNA Assays ›
Gene Symbol:
ZHX3
Gene Name:
zinc fingers and homeoboxes 3
Gene Aliases:
TIX1
Chromosome Location:
Chr.20: 41178449 - 41317606 on Build GRCh38
UniGene ID:
Hs.380133
Species:
Human
siRNA ID
HSS118099
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZHX3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_015035.3
NP_055850.1
3
1856
XM_005260341.3
XP_005260398.1
3
1883
XM_005260343.4
XP_005260400.1
3
1883
XM_006723748.3
XP_006723811.1
4
1944
XM_006723749.3
XP_006723812.1
2
1767
XM_011528701.2
XP_011527003.1
3
1820
XM_011528702.2
XP_011527004.1
6
2136
XM_011528703.2
XP_011527005.1
5
2088
XM_011528704.2
XP_011527006.1
3
1883
XM_011528705.2
XP_011527007.1
3
1899
XM_011528706.2
XP_011527008.1
3
1819
XM_011528707.2
XP_011527009.1
3
1872
XM_011528708.2
XP_011527010.1
3
1742
XM_011528709.2
XP_011527011.1
3
1744
XM_011528711.2
XP_011527013.1
3
1891
XM_011528713.2
XP_011527015.1
4
1981
XM_011528714.2
XP_011527016.1
4
1920
XM_011528717.2
XP_011527019.1
4
3595
XM_011528718.2
XP_011527020.1
4
1894
XM_011528720.1
XP_011527022.1
3
1891
XM_017027736.1
XP_016883225.1
3
1797
XM_017027737.1
XP_016883226.1
3
1899
XM_017027738.1
XP_016883227.1
3
2022
GenBank mRNA
AB007855.3
-
3
1827
AB081948.1
-
3
1838
AK090560.1
-
1
632
AK292415.1
-
3
1854
BC068569.1
-
4
1941
5295555
Back To Top
More Information
Panther Classification:
Molecular Function -
homeodomain transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
cell differentiation
positive regulation of osteoblast differentiation
negative regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription corepressor activity
protein binding
protein homodimerization activity
metal ion binding
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0