Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015079.5 | NP_055894.6 | 7 | 1566 | ||
NM_144572.1 | NP_653173.1 | 7 | 1566 | |||
XM_017022022.1 | XP_016877511.1 | 9 | 1587 | |||
GenBank mRNA | AB028978.1 | - | 7 | 1429 | ||
BC033712.1 | - | 2 | 349 | |||
BC140766.1 | - | 6 | 1211 | 9021634 | ||
BC144482.1 | - | 6 | 1211 | 9053009 | ||
BC152465.1 | - | 7 | 1429 | 8862601 |