Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZZEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015113.3 | NP_055928.3 | 46 | 7519 | ||
XM_005256561.2 | XP_005256618.1 | 46 | 7583 | |||
XM_017024382.1 | XP_016879871.1 | 46 | 7583 | |||
XM_017024383.1 | XP_016879872.1 | 46 | 7580 | |||
XM_017024384.1 | XP_016879873.1 | 21 | 3394 | |||
GenBank mRNA | AB007859.4 | - | 46 | 7519 | ||
BC067099.1 | - | 3 | 277 | |||
BC151836.1 | - | 46 | 7519 | 8860131 |