Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136115.2 | NP_001129587.1 | 4 | 874 | ||
NM_015176.3 | NP_055991.1 | 5 | 1070 | |||
NR_049764.1 | 4 | 931 | ||||
GenBank mRNA | AB007952.1 | - | 5 | 821 | ||
AK001628.1 | - | 5 | 1046 | |||
AK303381.1 | - | 4 | 899 | |||
AK307892.1 | - | 4 | 846 | |||
BC031576.1 | - | 5 | 1039 | 5188219 |