Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015176.3 | NP_055991.1 | 3, 4 | 560 | ||
GenBank mRNA | AB007952.1 | - | 3, 4 | 311 | ||
AK001628.1 | - | 3, 4 | 536 | |||
BC031576.1 | - | 3, 4 | 529 | 5188219 |