Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145114.1 | NP_001138586.1 | 20 | 2470 | ||
NM_001284337.1 | NP_001271266.1 | 19 | 2353 | |||
NM_015179.3 | NP_055994.2 | 22 | 2653 | |||
XM_011539555.1 | XP_011537857.1 | 21 | 2522 | |||
XM_011539556.2 | XP_011537858.1 | 22 | 2623 | |||
GenBank mRNA | AB014590.1 | - | 19 | 2266 | ||
AK021460.1 | - | 1370 | ||||
AK022496.1 | - | 22 | 2624 | |||
AK131283.1 | - | 16 | 1729 | |||
AK296315.1 | - | 20 | 2334 | |||
AL832924.1 | - | 5 | 1499 | |||
BC012745.2 | - | 22 | 2591 | 4025242 |