Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015191.2 | NP_056006.1 | 7, 8 | 1129 | ||
XM_017017417.1 | XP_016872906.1 | 7, 8 | 1088 | |||
XM_017017418.1 | XP_016872907.1 | 5, 6 | 658 | |||
GenBank mRNA | AB018324.3 | - | 7, 8 | 1030 | ||
AB084424.1 | - | 7, 8 | 950 | |||
AB096248.1 | - | 7, 8 | 1035 | |||
AK000396.1 | - | 7, 8 | 1035 | |||
AK291233.1 | - | 7, 8 | 1064 | |||
AL136764.2 | - | 7, 8 | 1052 | |||
BC078150.1 | - | 2, 3 | 312 | |||
BC113459.1 | - | 7, 8 | 986 | 8322511 | ||
BC117183.1 | - | 7, 8 | 986 | 40125734 |