Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EHBP1 SIRNA Assays ›
Gene Symbol:
EHBP1
Gene Name:
EH domain binding protein 1
Gene Aliases:
HPC12, NACSIN
Chromosome Location:
Chr.2: 62673851 - 63046487 on Build GRCh38
UniGene ID:
Hs.271667
Species:
Human
siRNA ID
HSS118550
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EHBP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142614.1
NP_001136086.1
12
2261
NM_001142615.2
NP_001136087.1
13
2494
NM_001142616.1
NP_001136088.1
13
2448
NM_015252.3
NP_056067.2
14
2553
XM_005264225.2
XP_005264282.1
14
2451
XM_005264226.2
XP_005264283.1
14
2808
XM_005264227.1
XP_005264284.1
13
2407
XM_006711971.2
XP_006712034.1
14
2512
XM_011532713.2
XP_011531015.1
14
2594
XM_011532714.2
XP_011531016.1
14
2443
XM_011532715.2
XP_011531017.1
14
2419
XM_011532716.2
XP_011531018.1
14
2512
XM_011532717.1
XP_011531019.1
14
2512
XM_011532718.2
XP_011531020.1
13
2173
XM_017003643.1
XP_016859132.1
14
2409
XM_017003644.1
XP_016859133.1
14
2386
XM_017003645.1
XP_016859134.1
14
2451
XM_017003646.1
XP_016859135.1
14
2514
XM_017003647.1
XP_016859136.1
13
2346
XM_017003648.1
XP_016859137.1
13
2342
XM_017003649.1
XP_016859138.1
13
2421
XM_017003650.1
XP_016859139.1
14
2451
XM_017003651.1
XP_016859140.1
13
2346
XM_017003652.1
XP_016859141.1
13
2407
XM_017003653.1
XP_016859142.1
13
2335
XM_017003654.1
XP_016859143.1
14
2512
GenBank mRNA
AB020710.1
-
6
1266
AK292545.1
-
13
2494
AL833968.1
-
14
2419
AY331186.1
-
14
2448
AY531390.1
-
12
1966
BC067215.1
-
13
2302
6502257
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0