Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278731.1 | NP_001265660.1 | 2 | 346 | ||
NM_001278732.1 | NP_001265661.1 | 3 | 521 | |||
NM_012208.3 | NP_036340.1 | 3 | 421 | |||
XM_011537619.1 | XP_011535921.1 | 4 | 434 | |||
XM_011537620.1 | XP_011535922.1 | 4 | 434 | |||
XM_017009287.1 | XP_016864776.1 | 3 | 421 | |||
XM_017009288.1 | XP_016864777.1 | 4 | 485 | |||
XM_017009289.1 | XP_016864778.1 | 4 | 478 | |||
XM_017009290.1 | XP_016864779.1 | 3 | 471 | |||
XM_017009291.1 | XP_016864780.1 | 3 | 164 | |||
XM_017009292.1 | XP_016864781.1 | 3 | 108 | |||
GenBank mRNA | AK293269.1 | - | 3 | 301 | ||
AK293390.1 | - | 2 | 163 | |||
AK309832.1 | - | 3 | 519 | |||
AK311277.1 | - | 3 | 517 | |||
AK312976.1 | - | 3 | 239 | |||
BC007680.2 | - | 3 | 240 | 3628006 | ||
BC014982.1 | - | 3 | 385 | 4859849 | ||
BQ228600.1 | - | 3 | 255 | |||
DA150729.1 | - | 3 | 423 | |||
U18937.1 | - | 3 | 347 |