Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317804.1 | NP_001304733.1 | 11 | 1199 | ||
NM_001317805.1 | NP_001304734.1 | 10 | 1714 | |||
NM_014294.5 | NP_055109.1 | 10 | 1282 | |||
XM_017013266.1 | XP_016868755.1 | 9 | 1154 | |||
XM_017013267.1 | XP_016868756.1 | 9 | 1498 | |||
GenBank mRNA | AK289397.1 | - | 10 | 1142 | ||
AK292777.1 | - | 10 | 1066 | |||
AK303411.1 | - | 11 | 1127 | |||
BC000687.2 | - | 10 | 1104 | 3347823 | ||
BC020283.1 | - | 10 | 1097 | |||
BC037738.1 | - | 10 | 1194 | 5261490 | ||
BT007359.1 | - | 10 | 1022 | |||
CR541654.1 | - | 10 | 1022 | |||
CR541739.1 | - | 10 | 1022 | |||
X63679.1 | - | 10 | 1143 |