Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAT6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256468.1 | NP_001243397.1 | 18 | 5850 | ||
NM_001256469.1 | NP_001243398.1 | 18 | 5523 | |||
NM_012330.3 | NP_036462.2 | 18 | 6399 | |||
XM_005269664.2 | XP_005269721.1 | 18 | 6460 | |||
XM_017016000.1 | XP_016871489.1 | 18 | 6308 | |||
XM_017016001.1 | XP_016871490.1 | 18 | 6172 | |||
XM_017016002.1 | XP_016871491.1 | 18 | 6411 | |||
XM_017016003.1 | XP_016871492.1 | 17 | 6082 | |||
XM_017016004.1 | XP_016871493.1 | 17 | 6146 | |||
XM_017016005.1 | XP_016871494.1 | 18 | 5759 | |||
XM_017016006.1 | XP_016871495.1 | 18 | 5432 | |||
XM_017016007.1 | XP_016871496.1 | 18 | 5295 | |||
XM_017016008.1 | XP_016871497.1 | 17 | 5206 | |||
XM_017016009.1 | XP_016871498.1 | 17 | 5361 | |||
GenBank mRNA | AB002381.2 | - | 16 | 4822 | ||
AF113514.1 | - | 17 | 5137 | |||
AF119230.1 | - | 17 | 5464 | |||
AF119231.1 | - | 17 | 6013 | |||
AF217500.1 | - | 18 | 6183 | |||
BC021128.1 | - | 2 | 2141 | |||
BC150270.1 | - | 16 | 4849 | 9007238 | ||
BC150618.1 | - | 18 | 6159 | 9056988 |