Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNPO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191028.2 | NP_001177957.2 | 14 | 2118 | ||
NM_012470.3 | NP_036602.1 | 15 | 2310 | |||
NR_034053.2 | 16 | 2471 | ||||
XM_011515989.2 | XP_011514291.1 | 15 | 1912 | |||
GenBank mRNA | AF145029.1 | - | 15 | 2122 | ||
AJ133769.1 | - | 15 | 1907 | |||
AK001263.1 | - | 8 | 864 | |||
AK022910.1 | - | 6 | 644 | |||
AK225999.1 | - | 14 | 2089 | |||
AK312580.1 | - | 15 | 2037 | |||
BC009221.2 | - | 3 | 154 | |||
BC068530.1 | - | 15 | 2038 |