Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LILRA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012276.4 | NP_036408.4 | 3 | 374 | ||
GenBank mRNA | AF041261.1 | - | 2 | 298 | ||
AF283989.1 | - | 1 | 197 | |||
BC109198.1 | - | 2 | 478 | 40005733 | ||
BC109199.1 | - | 3 | 320 | 40005735 | ||
BI834924.1 | - | 3 | 354 | |||
DA983933.1 | - | 3 | 374 |