Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM50A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014313.3 | NP_055128.1 | 3 | 321 | ||
XM_005245817.1 | XP_005245874.1 | 3 | 285 | |||
XM_011541159.1 | XP_011539461.1 | 3 | 515 | |||
GenBank mRNA | AF081282.1 | - | 3 | 248 | ||
AF087880.1 | - | 3 | 309 | |||
AK299180.1 | - | 3 | 285 | |||
AL136627.1 | - | 3 | 303 | |||
AY071927.1 | - | 3 | 298 | |||
AY358650.1 | - | 3 | 284 | |||
BC007341.2 | - | 3 | 269 | 3688672 | ||
BC066908.1 | - | 3 | 260 | 4817784 | ||
JX644905.1 | - | 3 | 321 | |||
JX644906.1 | - | 3 | 321 |