Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZIM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146326.1 | NP_001139798.1 | 10 | 1024 | ||
NM_001146327.1 | NP_001139799.1 | 10 | 1111 | |||
NM_015363.4 | NP_056178.3 | 9 | 948 | |||
GenBank mRNA | AF166122.1 | - | 9 | 931 |