Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BACE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207048.1 | NP_001193977.1 | 6 | 855 | ||
NM_001207049.1 | NP_001193978.1 | 6 | 780 | |||
NM_012104.4 | NP_036236.1 | 7 | 1493 | |||
NM_138971.3 | NP_620427.1 | 7 | 1361 | |||
NM_138972.3 | NP_620428.1 | 7 | 1418 | |||
NM_138973.3 | NP_620429.1 | 7 | 1286 | |||
GenBank mRNA | AB032975.2 | - | 7 | 1475 | ||
AB050436.1 | - | 7 | 960 | |||
AB050437.1 | - | 7 | 903 | |||
AB050438.1 | - | 7 | 828 | |||
AB177847.1 | - | 6 | 717 | |||
AF161367.1 | - | 3 | 321 | |||
AF190725.1 | - | 7 | 1485 | |||
AF200193.1 | - | 7 | 993 | |||
AF200343.1 | - | 7 | 1032 | |||
AF201468.1 | - | 7 | 1486 | |||
AF204943.1 | - | 7 | 1032 | |||
AF338816.1 | - | 7 | 957 | |||
AF338817.1 | - | 7 | 900 | |||
AK075049.1 | - | 7 | 968 | |||
AK296015.1 | - | 6 | 683 | |||
AK296535.1 | - | 6 | 768 | |||
BC036084.1 | - | 7 | 1488 | 5311572 | ||
BC065492.1 | - | 7 | 1493 | 6538154 |