Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R15A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014330.3 | NP_055145.3 | 2 | 464 | ||
GenBank mRNA | AK001361.1 | - | 2 | 435 | ||
AK225632.1 | - | 2 | 435 | |||
AK297615.1 | - | 3 | 352 | |||
BC003067.2 | - | 2 | 417 | 3507556 | ||
CR457259.1 | - | 1 | 195 | |||
DA484808.1 | - | 2 | 464 | |||
U83981.1 | - | 2 | 417 |