Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031696.3 | NP_001026866.1 | 5 | 587 | ||
NM_001291311.1 | NP_001278240.1 | 5 | 521 | |||
NM_012268.3 | NP_036400.2 | 5 | 811 | |||
XM_005258704.1 | XP_005258761.1 | 5 | 732 | |||
XM_005258707.4 | XP_005258764.1 | 5 | 514 | |||
XM_005258708.3 | XP_005258765.1 | 4 | 407 | |||
XM_005258709.4 | XP_005258766.1 | 4 | 395 | |||
XM_005258710.4 | XP_005258767.1 | 4 | 436 | |||
XM_006723122.1 | XP_006723185.1 | 5 | 899 | |||
XM_011526692.1 | XP_011524994.1 | 5 | 894 | |||
XM_011526693.1 | XP_011524995.1 | 5 | 614 | |||
XM_017026546.1 | XP_016882035.1 | 6 | 830 | |||
XM_017026547.1 | XP_016882036.1 | 5 | 398 | |||
XM_017026548.1 | XP_016882037.1 | 5 | 549 | |||
XM_017026549.1 | XP_016882038.1 | 5 | 652 | |||
GenBank mRNA | AK293690.1 | - | 4 | 312 | ||
AY927565.1 | - | 2 | 323 | |||
BC000553.1 | - | 1 | 57 | |||
BC036327.1 | - | 5 | 312 | 5189261 | ||
BC096820.1 | - | 5 | 510 | 30558682 | ||
BP304863.1 | - | 4 | 327 | |||
U60644.1 | - | 5 | 517 |