Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other OSBP2 SIRNA Assays ›
Gene Symbol:
OSBP2
Gene Name:
oxysterol binding protein 2
Gene Aliases:
HLM, ORP-4, ORP4, OSBPL1, OSBPL4
Chromosome Location:
Chr.22: 30693782 - 30907824 on Build GRCh38
UniGene ID:
Hs.517546
Species:
Human
siRNA ID
HSS119193
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: OSBP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282738.1
NP_001269667.1
4
709
NM_001282739.1
NP_001269668.1
3
1029
NM_001282740.1
NP_001269669.1
2
212
NM_001282741.1
NP_001269670.1
2
266
NM_030758.3
NP_110385.1
3
1029
XM_005261465.4
XP_005261522.1
3
955
XM_006724203.2
XP_006724266.2
3
955
XM_006724207.2
XP_006724270.1
2
212
XM_011530057.1
XP_011528359.1
3
955
XM_011530058.1
XP_011528360.1
3
955
XM_011530059.2
XP_011528361.1
3
955
XM_011530060.1
XP_011528362.1
2
266
XM_011530062.1
XP_011528364.1
3
311
XM_017028708.1
XP_016884197.1
3
955
XM_017028709.1
XP_016884198.1
3
955
XM_017028710.1
XP_016884199.1
3
418
XM_017028711.1
XP_016884200.1
2
716
XM_017028712.1
XP_016884201.1
2
216
XM_017028713.1
XP_016884202.1
2
542
GenBank mRNA
AB051451.2
-
3
937
AF288741.1
-
3
923
AF323731.1
-
3
1029
AK093196.1
-
2
212
AK097581.1
-
3
961
AK131374.1
-
3
556
AK294007.1
-
3
961
AK296354.1
-
2
266
AK296525.1
-
4
711
BC118914.2
-
3
946
40037933
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
lipid transport
spermatid development
protein binding
cholesterol binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0