Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL2L13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270726.1 | NP_001257655.1 | 4, 5 | 628 | ||
NM_001270729.1 | NP_001257658.1 | 4, 5 | 642 | |||
NM_001270730.1 | NP_001257659.1 | 3, 4 | 471 | |||
NM_015367.3 | NP_056182.2 | 5, 6 | 799 | |||
XM_011546119.1 | XP_011544421.1 | 5, 6 | 1162 | |||
XM_011546120.1 | XP_011544422.1 | 5, 6 | 1164 | |||
XM_011546121.2 | XP_011544423.1 | 6, 7 | 1147 | |||
XM_017028726.1 | XP_016884215.1 | 7, 8 | 1448 | |||
XM_017028728.1 | XP_016884217.1 | 5, 6 | 1176 | |||
XM_017028731.1 | XP_016884220.1 | 4, 5 | 1005 | |||
XM_017028733.1 | XP_016884222.1 | 5, 6 | 799 | |||
GenBank mRNA | AF146568.1 | - | 2, 3 | 220 | ||
AF246665.1 | - | 6, 7 | 1185 | |||
AF325209.1 | - | 4, 5 | 446 | |||
AK056248.1 | - | 4, 5 | 485 | |||
AK314489.1 | - | 5, 6 | 626 | |||
BC007658.2 | - | 5, 6 | 558 | 3343994 | ||
BX354578.2 | - | 4, 5 | 507 | |||
BX407004.2 | - | 4, 5 | 381 | |||
CR456521.1 | - | 5, 6 | 702 | |||
DA154984.1 | - | 3, 4 | 298 |