Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPDEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012391.2 | NP_036523.1 | 4, 5 | 1113 | ||
XM_005248988.4 | XP_005249045.2 | 4, 5 | 1805 | |||
GenBank mRNA | AB031549.1 | - | 4, 5 | 1084 | ||
AF071538.1 | - | 4, 5 | 1094 | |||
BC021299.2 | - | 4, 5 | 1090 | 4546860 |