Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014033.3 | NP_054752.3 | 2 | 801 | ||
XM_006719332.1 | XP_006719395.1 | 3 | 1155 | |||
GenBank mRNA | AF113007.1 | - | 2 | 475 | ||
AJ536233.1 | - | 2 | 523 | |||
AK023693.1 | - | 2 | 533 | |||
AK024409.1 | - | 2 | 566 | |||
AK225595.1 | - | 2 | 538 | |||
AK293356.1 | - | 3 | 327 | |||
AK307071.1 | - | 3 | 393 | |||
AL050159.1 | - | 2 | 511 | |||
AY358797.1 | - | 2 | 569 | |||
BC004492.2 | - | 2 | 518 | 3689692 | ||
BC008180.2 | - | 2 | 519 | 2900478 | ||
BM805242.1 | - | 2 | 825 |