Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCAF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015397.3 | NP_056212.1 | 2 | 537 | ||
XM_011517843.1 | XP_011516145.1 | 2 | 364 | |||
GenBank mRNA | AB067479.1 | - | 2 | 457 | ||
AK000354.1 | - | 2 | 315 | |||
AK292935.1 | - | 2 | 343 | |||
BC008893.2 | - | 1 | 100 | |||
BC063823.1 | - | 2 | 455 | 6184796 | ||
BG718613.1 | - | 2 | 508 | |||
DQ518909.1 | - | 2 | 457 |