Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE4D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165899.1 | NP_001159371.1 | 1 | 129 | ||
XM_017009566.1 | XP_016865055.1 | 1 | 96 | |||
XM_017009568.1 | XP_016865057.1 | 1 | 56 | |||
XM_017009571.1 | XP_016865060.1 | 1 | 52 | |||
GenBank mRNA | AK307363.1 | - | 1 | 96 | ||
AY245866.1 | - | 1 | 94 | |||
DB230503.1 | - | 1 | 129 |
Gene Symbol: PART1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024617.1 | 1 | 254 | |||
NR_028508.1 | 1 | 35 | ||||
NR_028509.1 | 1 | 35 | ||||
GenBank mRNA | AF163475.1 | - | 1 | 35 | ||
AK312071.1 | - | 1 | 257 |