Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WHRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083885.2 | NP_001077354.2 | 3 | 391 | ||
NM_001173425.1 | NP_001166896.1 | 3 | 1555 | |||
NM_015404.3 | NP_056219.3 | 3 | 1555 | |||
XM_011518484.1 | XP_011516786.1 | 3 | 1608 | |||
XM_011518485.1 | XP_011516787.1 | 4 | 1456 | |||
XM_011518486.1 | XP_011516788.1 | 3 | 1608 | |||
XM_011518488.1 | XP_011516790.1 | 3 | 1608 | |||
XM_011518489.2 | XP_011516791.1 | 3 | 1608 | |||
XM_011518491.2 | XP_011516793.1 | 3 | 1608 | |||
XM_011518492.1 | XP_011516794.1 | 3 | 1608 | |||
XM_011518494.2 | XP_011516796.1 | 3 | 1608 | |||
XM_011518495.1 | XP_011516797.1 | 1 | 63 | |||
GenBank mRNA | AB040959.1 | - | 3 | 1069 | ||
AL110228.1 | - | 3 | 349 | |||
BC014524.1 | - | 2055 | ||||
BC136416.1 | - | 3 | 1304 | 9020404 | ||
BC142614.1 | - | 3 | 1555 | 40147287 | ||
BC142684.1 | - | 3 | 1530 | 40148465 | ||
DA502065.1 | - | 3 | 391 |