Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEPRO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319109.1 | NP_001306038.1 | 7 | 1545 | ||
NM_001319110.1 | NP_001306039.1 | 6 | 1390 | |||
NM_001319111.1 | NP_001306040.1 | 8 | 1359 | |||
NM_001319112.1 | NP_001306041.1 | 8 | 1558 | |||
NM_001319114.1 | NP_001306043.1 | 8 | 1604 | |||
NM_001319115.1 | NP_001306044.1 | 8 | 1600 | |||
NM_015412.3 | NP_056227.2 | 9 | 1730 | |||
NR_134969.1 | 6 | 1483 | ||||
XM_005247276.2 | XP_005247333.1 | 9 | 1697 | |||
XM_011512635.2 | XP_011510937.1 | 9 | 1452 | |||
XM_011512636.2 | XP_011510938.1 | 9 | 1456 | |||
XM_011512638.2 | XP_011510940.1 | 8 | 1351 | |||
XM_017006102.1 | XP_016861591.1 | 9 | 1719 | |||
GenBank mRNA | AK024325.1 | - | 9 | 1688 | ||
AK295165.1 | - | 8 | 1489 | |||
AK295954.1 | - | 6 | 1327 | |||
AK300811.1 | - | 8 | 1292 | |||
AK301161.1 | - | 8 | 1518 | |||
AK304022.1 | - | 3 | 818 | |||
AK304083.1 | - | 8 | 1533 | |||
AL117573.1 | - | 9 | 1672 | |||
BC014673.2 | - | 8 | 2188 | |||
BC067743.1 | - | 9 | 1690 | 4812681 | ||
CR749341.1 | - | 7 | 1546 |