Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015417.4 | NP_056232.2 | 2 | 289 | ||
XM_005260683.4 | XP_005260740.1 | 2 | 300 | |||
GenBank mRNA | AK313766.1 | - | 2 | 302 | ||
AL080154.1 | - | 2 | 271 | |||
BC022476.1 | - | 2 | 248 | 4816764 | ||
BI520334.1 | - | 2 | 288 | |||
DB030684.1 | - | 2 | 302 | |||
DB033051.1 | - | 2 | 282 | |||
EF560745.1 | - | 2 | 287 |