Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129727.2 | NP_001123199.1 | 3 | 455 | ||
NM_001129728.1 | NP_001123200.1 | 2 | 415 | |||
NM_001129729.2 | NP_001123201.1 | 2 | 388 | |||
NM_001129731.2 | NP_001123203.1 | 1 | 637 | |||
XM_011522985.2 | XP_011521287.1 | 2 | 385 | |||
XM_011522986.2 | XP_011521288.1 | 2 | 517 | |||
XM_011522987.2 | XP_011521289.1 | 3 | 461 | |||
XM_011522988.2 | XP_011521290.1 | 2 | 334 | |||
GenBank mRNA | AB197663.1 | - | 1 | 2651 | ||
AB197664.1 | - | 1 | 2651 | |||
AK024475.1 | - | 2 | 350 | |||
AK308298.1 | - | 2 | 324 | |||
BC001520.2 | - | 2 | 329 | |||
BC054486.1 | - | 1 | 572 | 4820741 | ||
BC063501.1 | - | 2 | 326 | 5527705 | ||
BC082974.1 | - | 3 | 423 | 6291175 | ||
BC115390.1 | - | 1 | 265 | |||
DB061030.1 | - | 2 | 415 | |||
DB279205.1 | - | 3 | 455 | |||
DB461411.1 | - | 2 | 388 |