Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129727.2 | NP_001123199.1 | 6 | 1093 | ||
NM_001129728.1 | NP_001123200.1 | 5 | 1053 | |||
NM_001129729.2 | NP_001123201.1 | 5 | 1026 | |||
NM_001129731.2 | NP_001123203.1 | 3 | 1032 | |||
XM_011522985.2 | XP_011521287.1 | 5 | 1023 | |||
XM_011522986.2 | XP_011521288.1 | 5 | 1155 | |||
XM_011522987.2 | XP_011521289.1 | 6 | 1099 | |||
XM_011522988.2 | XP_011521290.1 | 5 | 972 | |||
GenBank mRNA | AB197663.1 | - | 4 | 3289 | ||
AB197664.1 | - | 3 | 3456 | |||
AK024475.1 | - | 5 | 988 | |||
AK304723.1 | - | 3 | 444 | |||
AK308298.1 | - | 5 | 962 | |||
BC001520.2 | - | 4 | 511 | |||
BC054486.1 | - | 3 | 967 | 4820741 | ||
BC063501.1 | - | 5 | 964 | 5527705 | ||
BC082974.1 | - | 6 | 1061 | 6291175 | ||
BC115390.1 | - | 3 | 1070 |