Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF19A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001280539.1 | NP_001267468.1 | 3 | 675 | ||
NM_015435.4 | NP_056250.3 | 3 | 706 | |||
NM_183419.3 | NP_904355.1 | 2 | 782 | |||
XM_005250853.3 | XP_005250910.1 | 2 | 744 | |||
XM_011516956.2 | XP_011515258.1 | 2 | 767 | |||
XM_017013302.1 | XP_016868791.1 | 3 | 867 | |||
XM_017013303.1 | XP_016868792.1 | 3 | 777 | |||
XM_017013304.1 | XP_016868793.1 | 2 | 724 | |||
XM_017013305.1 | XP_016868794.1 | 1 | 471 | |||
GenBank mRNA | AB029316.1 | - | 3 | 706 | ||
BC093938.1 | - | 1 | 478 | 7939783 | ||
BC093940.2 | - | 1 | 478 | 7939785 |