Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCHY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009922.2 | NP_001009922.1 | 7, 8 | 781 | ||
NM_001278536.1 | NP_001265465.1 | 7, 8 | 688 | |||
NM_001278537.1 | NP_001265466.1 | 6, 7 | 661 | |||
NM_001278538.1 | NP_001265467.1 | 8, 9 | 764 | |||
NM_001278539.1 | NP_001265468.1 | 8, 9 | 846 | |||
NM_015436.3 | NP_056251.2 | 8, 9 | 808 | |||
NR_037913.1 | 8, 9 | 836 | ||||
NR_037914.1 | 8, 9 | 832 | ||||
NR_103723.1 | 8, 9 | 806 | ||||
NR_103724.1 | 8, 9 | 770 | ||||
NR_103725.1 | 6, 7 | 555 | ||||
XM_011531838.2 | XP_011530140.1 | 8, 9 | 777 | |||
XM_011531839.2 | XP_011530141.1 | 8, 9 | 965 | |||
GenBank mRNA | AF247041.1 | - | 8, 9 | 664 | ||
AF255666.1 | - | 8, 9 | 690 | |||
AF305424.1 | - | 8, 9 | 698 | |||
AK091501.1 | - | 8, 9 | 779 | |||
AK308754.1 | - | 7, 8 | 541 | |||
AK308848.1 | - | 5, 6 | 590 | |||
AK309555.1 | - | 8, 9 | 679 | |||
AL050144.1 | - | 7, 8 | 565 | |||
AY888047.1 | - | 8, 9 | 652 | |||
BC015464.1 | - | 3, 4 | 233 | |||
BC047393.1 | - | 8, 9 | 808 | 5301408 | ||
BG703058.1 | - | 8, 9 | 677 | |||
BI913695.1 | - | 6, 7 | 473 | |||
BP314571.1 | - | 6, 7 | 528 | |||
BQ959587.1 | - | 8, 9 | 637 | |||
CF597260.1 | - | 6, 7 | 377 | |||
GQ250944.1 | - | 8, 9 | 664 | |||
GQ250945.1 | - | 7, 8 | 637 | |||
GU937000.1 | - | 8, 9 | 692 | |||
JF691538.1 | - | 8, 9 | 664 | |||
JF691539.1 | - | 8, 9 | 664 |