Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACOT11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015547.3 | NP_056362.1 | 5 | 585 | ||
NM_147161.3 | NP_671517.1 | 5 | 585 | |||
GenBank mRNA | AB014607.1 | - | 5 | 480 | ||
AF416921.1 | - | 5 | 427 | |||
AF416922.1 | - | 5 | 431 | |||
AK023937.1 | - | 5 | 491 | |||
AK091112.1 | - | 4 | 487 | |||
AK223570.1 | - | 5 | 491 | |||
BC001517.2 | - | 6 | 652 | |||
BC093844.1 | - | 5 | 451 | 7939689 | ||
BC093846.1 | - | 5 | 451 | 7939691 |