Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IPCEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130699.1 | NP_001124171.1 | 6 | 372 | ||
NM_001130700.1 | NP_001124172.1 | 5 | 332 | |||
NM_015553.2 | NP_056368.1 | 5 | 329 | |||
XM_005266919.4 | XP_005266976.1 | 6 | 489 | |||
XM_005266921.4 | XP_005266978.1 | 5 | 415 | |||
XM_005266923.3 | XP_005266980.1 | 6 | 423 | |||
XM_011535742.2 | XP_011534044.1 | 6 | 487 | |||
XM_011535744.2 | XP_011534046.1 | 2 | 232 | |||
XM_017010716.1 | XP_016866205.1 | 6 | 459 | |||
XM_017010717.1 | XP_016866206.1 | 5 | 573 | |||
XM_017010718.1 | XP_016866207.1 | 5 | 570 | |||
GenBank mRNA | AB007863.1 | - | 1 | 110 | ||
AJ310566.1 | - | 5 | 330 | |||
AK289917.1 | - | 5 | 301 | |||
BC105015.1 | - | 4 | 238 | 8144018 | ||
BC105043.1 | - | 4 | 238 | 8144046 | ||
BC143623.1 | - | 4 | 241 | 9052136 | ||
BC143629.1 | - | 4 | 238 | 9052142 | ||
BX647254.1 | - | 6 | 372 |