Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCTN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143973.1 | NP_001137445.1 | 10 | 1507 | ||
NM_015631.5 | NP_056446.4 | 14 | 1951 | |||
XM_005269690.1 | XP_005269747.1 | 13 | 1712 | |||
XM_011539627.1 | XP_011537929.1 | 14 | 1778 | |||
XM_011539628.1 | XP_011537930.1 | 13 | 1655 | |||
GenBank mRNA | AK075357.1 | - | 14 | 1737 | ||
AK098295.1 | - | 12 | 1848 | |||
AK128834.1 | - | 13 | 2748 | |||
AK299141.1 | - | 10 | 1291 | |||
AL050022.1 | - | 14 | 1735 | |||
AY359022.1 | - | 14 | 1715 | |||
BC009494.2 | - | 11 | 1111 | |||
BC068449.1 | - | 14 | 1951 | 5298627 | ||
DQ278872.1 | - | 14 | 1951 |