Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C10orf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015652.2 | NP_056467.2 | 1 | 2005 | ||
GenBank mRNA | AK023084.1 | - | 1 | 1194 | ||
AK126886.1 | - | 1 | 2058 | |||
BC024315.1 | - | 1 | 2005 | 3903728 | ||
BC031482.1 | - | 1 | 1402 |