Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSL1D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015659.2 | NP_056474.2 | 2, 3 | 322 | ||
GenBank mRNA | AJ007398.1 | - | 2, 3 | 254 | ||
AK000822.1 | - | 2, 3 | 266 | |||
AL049999.1 | - | 2, 3 | 309 | |||
AY154473.1 | - | 2, 3 | 283 | |||
AY598331.1 | - | 2, 3 | 249 | |||
BC016362.1 | - | 2, 3 | 285 | |||
BC017899.1 | - | 2, 3 | 271 | |||
BC019069.2 | - | 2, 3 | 255 | |||
BC070325.1 | - | 2, 3 | 273 | |||
BC107782.1 | - | 2, 3 | 275 | |||
BC112228.1 | - | 2, 3 | 287 | 8327696 | ||
BC113699.1 | - | 2, 3 | 275 | 8322751 | ||
BI856275.1 | - | 2, 3 | 322 |