Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193534.1 | NP_001180463.1 | 10 | 2004 | ||
NM_001193535.1 | NP_001180464.1 | 10 | 1956 | |||
NM_012161.3 | NP_036293.1 | 10 | 2007 | |||
NR_036464.1 | 11 | 2111 | ||||
XM_006713959.3 | XP_006714022.1 | 11 | 2103 | |||
XM_011513831.2 | XP_011512133.1 | 10 | 1789 | |||
XM_011513832.2 | XP_011512134.1 | 10 | 1936 | |||
XM_011513833.2 | XP_011512135.1 | 9 | 1747 | |||
XM_017008019.1 | XP_016863508.1 | 12 | 2874 | |||
GenBank mRNA | AF052097.1 | - | 2 | 566 | ||
AF142481.1 | - | 10 | 1929 | |||
AF157323.1 | - | 10 | 1868 | |||
AF174591.1 | - | 7 | 1414 | |||
AF176700.1 | - | 10 | 1891 | |||
AF199420.1 | - | 10 | 1893 | |||
AK000153.1 | - | 9 | 1717 | |||
AK295510.1 | - | 10 | 1926 | |||
AK310860.1 | - | 10 | 1936 | |||
AL832808.1 | - | 8 | 1643 | |||
BC012320.1 | - | 11 | 2040 | |||
BC030656.1 | - | 10 | 1978 | 4837080 | ||
BX537957.1 | - | 10 | 1902 |