Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278716.1 | NP_001265645.1 | 4 | 715 | ||
NM_012160.4 | NP_036292.2 | 3 | 597 | |||
NR_103836.1 | 3 | 597 | ||||
NR_103837.1 | 3 | 597 | ||||
XM_005266930.2 | XP_005266987.1 | 4 | 703 | |||
XM_011535748.2 | XP_011534050.1 | 4 | 703 | |||
XM_017010726.1 | XP_016866215.1 | 3 | 529 | |||
XM_017010727.1 | XP_016866216.1 | 3 | 557 | |||
XM_017010728.1 | XP_016866217.1 | 4 | 715 | |||
GenBank mRNA | AF174590.1 | - | 1 | 206 | ||
AF176699.1 | - | 1 | 206 | |||
AF199355.1 | - | 1 | 210 | |||
AK313076.1 | - | 3 | 526 | |||
BC055010.1 | - | 3 | 536 | 6137889 | ||
BC091484.1 | - | 4 | 635 | 6178874 | ||
BQ213738.1 | - | 3 | 577 |