Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257194.1 | NP_001244123.1 | 2, 3 | 742 | ||
NM_017415.2 | NP_059111.2 | 2, 3 | 567 | |||
GenBank mRNA | AB032955.1 | - | 2, 3 | 337 | ||
AF208068.1 | - | 2, 3 | 251 | |||
AF208070.1 | - | 2, 3 | 742 | |||
AK314707.1 | - | 2, 3 | 308 | |||
BC034035.1 | - | 2, 3 | 566 | 4817650 |