Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033506.2 | NP_277041.1 | 1 | 307 | ||
GenBank mRNA | AF174604.1 | - | 1 | 14 | ||
AK292240.1 | - | 1 | 314 | |||
AK310487.1 | - | 1 | 252 | |||
AL136811.1 | - | 1 | 252 | |||
BC067293.1 | - | 1 | 247 | 30383439 |