Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL1RAPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017416.1 | NP_059112.1 | 11 | 2329 | ||
XM_011530905.2 | XP_011529207.1 | 9 | 1337 | |||
XM_017029403.1 | XP_016884892.1 | 11 | 1747 | |||
GenBank mRNA | AF181285.1 | - | 9 | 1491 | ||
AF212016.1 | - | 10 | 1573 | |||
AF284436.1 | - | 11 | 1724 | |||
AJ272208.1 | - | 11 | 2329 | |||
AJ290436.1 | - | 10 | 1592 | |||
AK290196.1 | - | 11 | 2445 | |||
AK297237.1 | - | 3 | 625 | |||
BC104784.1 | - | 11 | 1621 | 8143787 | ||
BC104786.1 | - | 11 | 1621 | 8143789 |